Learn More
Bellco Glass, Inc. 100mm x 20mm Petri Dish w/Grip Ring, Sterile
SDP
Catalog No. 502330560
Encompass_Preferred
Non-treated Cell Culture (Petri) Dishes have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications60, 70 and 100mm dishes available with a pronounced grip ring for improved handlingVented lids contain molded spacers to allow gas exchange Pack Description10/BagCase Quantity300SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume15-16 mLTissue Culture TreatedNoCell Growth Area55cm²RNase/DNase FreeYes
Catalog No. 50-233-0560 Supplier Bellco Glass, Inc. Supplier No. 229623000
Non-treated Cell Culture (Petri) Dishes have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications60, 70 and 100mm dishes available with a pronounced grip ring for improved handlingVented lids contain molded spacers to allow gas exchange Pack Description10/BagCase Quantity300SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume15-16 mLTissue Culture TreatedNoCell Growth Area55cm²RNase/DNase FreeYes