Learn More
Bellco Glass, Inc. 100mm x 20mm Tissue Culture
SDP
Catalog No. 502330195
Encompass_Preferred
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions.  60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid Working Volume 1516mL Working Area 60.5cm2 Case Quantity 300
Catalog No. 50-233-0195 Supplier Bellco Glass, Inc. Supplier No. 229621000
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions.  60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid Working Volume 1516mL Working Area 60.5cm2 Case Quantity 300