Learn More
Agilent Technologies 100MU/40UL GLYKO N-GLYCANASE
Catalog No. NC9039160
Encompass
PNGase F, peptide-N-glycosidase F,peptide-N4-(N-acetyl--glucosaminyl)asparagine amidase.Releases intact N-glycans by cleavingbetween the innermost GlcNAc and Asn.
Catalog No. NC9039160 Supplier Agilent Technologies Supplier No. GKE5006A
PNGase F, peptide-N-glycosidase F,peptide-N4-(N-acetyl--glucosaminyl)asparagine amidase.Releases intact N-glycans by cleavingbetween the innermost GlcNAc and Asn.