Learn More
Nanocomposix Ultra Uniform Gold Nanoparticle Number Standard - 100 nm, 1E8 particles/mL, 25 mL
SDP
Catalog No. NC2885657
Encompass_Preferred
Ultra Uniform materials are our most highly-monodispersed particles. Ideal for use as size standards, applications requiring a tight size distribution. Our Ultra Uniform Particle Number Standards provide additional characterization and careful formulation for applications that require a known particle number concentration.PEG-carboxyl is covalently bound to the metal surface. The acid provides a highly negatively charged surface and can be utilized for conjugation.Provided at 10^8 particles/mL in aqueous 1 mM sodium citrate.APPLICATIONS- sp-ICP-MS: Single Particle Inductively Coupled Plasma Mass Spectroscopy- NTA: Nano Tracking Analysis- TRPS: Tunable Resistive Pulse Sensing
Catalog No. NC2885657 Supplier Nanocomposix Supplier No. AUXS10025M
Ultra Uniform materials are our most highly-monodispersed particles. Ideal for use as size standards, applications requiring a tight size distribution. Our Ultra Uniform Particle Number Standards provide additional characterization and careful formulation for applications that require a known particle number concentration.PEG-carboxyl is covalently bound to the metal surface. The acid provides a highly negatively charged surface and can be utilized for conjugation.Provided at 10^8 particles/mL in aqueous 1 mM sodium citrate.APPLICATIONS- sp-ICP-MS: Single Particle Inductively Coupled Plasma Mass Spectroscopy- NTA: Nano Tracking Analysis- TRPS: Tunable Resistive Pulse Sensing