Learn More
Nanocomposix NanoXact Gold Nanospheres - Bare (Citrate) - 100 nm, 0.05 mg/mL in aq. 2 mM sodium citrate, 50 mL
SDP
Catalog No. NC2885644
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.100 nm spherical gold nanoparticles combine strong plasmonic optical properties with versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from immunodiagnostics, biosensors, sensing, optical effects, drug delivery, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC2885644 Supplier Nanocomposix Supplier No. AUCN10050M
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.100 nm spherical gold nanoparticles combine strong plasmonic optical properties with versatile surface functionalization capabilities. These nanoparticles are useful in a broad range of applications from immunodiagnostics, biosensors, sensing, optical effects, drug delivery, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.