Learn More
USA Scientific Inc 100pl cardboard grid 4.8"x4.8"
Catalog No. 502745203
Encompass
100-place cardboard grid measures approximately 4.8inch W x 4.8inch L x 1.4inch H. For tubes up to 12mm inch diameter.
Catalog No. 50-274-5203 Supplier USA Scientific Inc Supplier No. 90230100
100-place cardboard grid measures approximately 4.8inch W x 4.8inch L x 1.4inch H. For tubes up to 12mm inch diameter.