Learn More
Organomation 100 Position MULTIVAP Nitrogen Evaporator
SDP
Catalog No. NC2124743
Encompass_Preferred
The 100 position MULTIVAP parallel evaporator is a water bath model. This instrument contains a rectangular gas distribution system mounted on a frame and dual post assembly. The manifold delivers nitrogen gas to the needles or glass pipettes, which directs the gas onto the sample’s surface. This method results in rapid solvent evaporation and sample concentration. In order to conserve nitrogen gas, the manifold has a toggle switch for every row of 10 samples, which allows individual rows to be shut off when evaporating batches of less than 100 samples. Nitrogen gas flow is also controlled by an adjustable flow meter and a built-in pressure regulator. The 100 position MULTIVAP small vial evaporator uses a heated water bath to reach evaporative temperatures ranging from 30°C to 100°C.
This item is not returnable. View return policy
Catalog No. NC2124743 Supplier Organomation Supplier No. 11300
Add to Cart
Add to Cart
Request a Quote
The 100 position MULTIVAP parallel evaporator is a water bath model. This instrument contains a rectangular gas distribution system mounted on a frame and dual post assembly. The manifold delivers nitrogen gas to the needles or glass pipettes, which directs the gas onto the sample’s surface. This method results in rapid solvent evaporation and sample concentration. In order to conserve nitrogen gas, the manifold has a toggle switch for every row of 10 samples, which allows individual rows to be shut off when evaporating batches of less than 100 samples. Nitrogen gas flow is also controlled by an adjustable flow meter and a built-in pressure regulator. The 100 position MULTIVAP small vial evaporator uses a heated water bath to reach evaporative temperatures ranging from 30°C to 100°C.