Learn More
Sensit Technologies Combo Calibration Gas, CH4, CO, 103L
SDP
Catalog No. NC1663365
Encompass
Calibration gas mixture of 100ppm Carbon Monoxide and 2.5% Methane (50% LEL) in Air. Cylinder has a 103L capacity, and is made of aluminum. Has a maximum pressure of 1000psi and an accuracy of ±2%. Has a 5/8"-18 UNF Threaded Collar connection. For use with the SENSIT Gold G2 Combustible Gas Leak Detector and Trak-It IIIa Combustible Gas Indicator.
Catalog No. NC1663365 Supplier Sensit Technologies Supplier No. 315140001
Calibration gas mixture of 100ppm Carbon Monoxide and 2.5% Methane (50% LEL) in Air. Cylinder has a 103L capacity, and is made of aluminum. Has a maximum pressure of 1000psi and an accuracy of ±2%. Has a 5/8"-18 UNF Threaded Collar connection. For use with the SENSIT Gold G2 Combustible Gas Leak Detector and Trak-It IIIa Combustible Gas Indicator.