Learn More
Optimize Technologies 100psi Back Pressure Regulator
SDP
Catalog No. 50113278
Encompass_Preferred
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates. Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100uL internal volume and includes fittings for immediate connection out of the box. Key Features include: a flow-through design that utilizes an active mechanism to maintain steady back pressure across a range of conditions, the product prevents solvent decompression and bubble formation in the flow cell, reduces baseline noise and drift, has a 100uL internal volume, and comes with OPTI-LOK™ LP Fittings.
Catalog No. 50-113-278 Supplier Optimize Technologies Supplier No. 100600130
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates. Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100uL internal volume and includes fittings for immediate connection out of the box. Key Features include: a flow-through design that utilizes an active mechanism to maintain steady back pressure across a range of conditions, the product prevents solvent decompression and bubble formation in the flow cell, reduces baseline noise and drift, has a 100uL internal volume, and comes with OPTI-LOK™ LP Fittings.