Learn More
Athens Research And Technology Inc Antithrombin III (ATIII, SERPINC1), Native Human Plasma, ≥95% (SDS-PAGE), 100 µg
Catalog No. 50489622
Encompass_Preferred
Antithrombin III (ATIII, SERPINC1) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 100 µg pack. Molecular weight: 58 kDa. Antithrombin III, Human Plasma: 100 UG; Lyophilized from 50 mM Tris-HCl, pH 8.0, with 150 mM NaCl; MW 58,000; EC=0.7; Purity: > =95% by SDS-PAGE;
Catalog No. 50-489-622 Supplier Athens Research And Technology Inc Supplier No. 1616012020100UG
Antithrombin III (ATIII, SERPINC1) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 100 µg pack. Molecular weight: 58 kDa. Antithrombin III, Human Plasma: 100 UG; Lyophilized from 50 mM Tris-HCl, pH 8.0, with 150 mM NaCl; MW 58,000; EC=0.7; Purity: > =95% by SDS-PAGE;