Learn More
Athens Research And Technology Inc 100UG APO CIII HUMAN PLASMA
Supplier:  Athens Research And Technology Inc 1616120303100UG
Encompass_Preferred
Apolipoprotein CIII, Human Plasma, VLDL: 100 UG; Frozen in 10 mM NH4HCO3, pH 7.4; MW 8,750; Purity: > =95% by SDS-PAGE; This native protein is prepared and purified from fresh, non-frozen human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-650