Learn More
Athens Research And Technology Inc 100UG AZUROCIDIN HU NEUTROPHIL
Supplier:  Athens Research And Technology Inc 1614012621100UG
Encompass_Preferred
Azurocidin, Human Neutrophil (Cationic Protein 37): 100 UG; Salt-free lyophilized solid; MW 37,000; EC= 0.51; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from whole blood in the Athens Research & Tech lab.
Catalog No. 50-489-668