Learn More
Advanced Targeting Systems 100UG FAB-ZAP HUMAN SECND CONJ
Catalog No. NC9066515
Encompass
Fab-ZAP is a chemical conjugate of goat anti-human monovalent antibody and the ribosome-inactivating protein, saporin.
Catalog No. NC9066515 Supplier Advanced Targeting Systems Supplier No. IT51100
Fab-ZAP is a chemical conjugate of goat anti-human monovalent antibody and the ribosome-inactivating protein, saporin.