Learn More
Inorganic Ventures 1000µg/mL IODIDE 125mL
SDP
Catalog No. NC0784866
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Iodide for ICP is a 125mL certified reference material set in a water matrix for stability. This iodide standard isTraceable to NISTand manufactured in accordance with our stringentQuality Assuranceguidelines. One yearExpiration Date. Can be used for analyzing Iodide by ICP-OES.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC0784866 Supplier Inorganic Ventures Supplier No. CGICI1125ML
Add to Cart
Add to Cart
Inorganic Ventures' 1000 ppm (µg/mL) Iodide for ICP is a 125mL certified reference material set in a water matrix for stability. This iodide standard isTraceable to NISTand manufactured in accordance with our stringentQuality Assuranceguidelines. One yearExpiration Date. Can be used for analyzing Iodide by ICP-OES.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search