Learn More
Inorganic Ventures 100µg/mL MERCURY HNO3 125mL
SDP
Catalog No. NC1798585
Encompass_Preferred
100ug/mL MERCURY HNO3 has a matrix of 10% v/v Nitric Acid and a starting material of Hg metal.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1798585 Supplier Inorganic Ventures Supplier No. MSHGN100PPM125ML
100ug/mL MERCURY HNO3 has a matrix of 10% v/v Nitric Acid and a starting material of Hg metal.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search