Learn More
Abcam Recombinant Human Calreticulin protein (His tag), 100UG
Supplier:  Abcam AB235685100UG
Encompass_Preferred
Recombinant Human Calreticulin protein (His tag) is a Human Full Length protein, in the 18 to 417 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: CRTC, CALR, Calreticulin, CRP55, Calregulin, Endoplasmic reticulum resident protein 60, HACBP, grp60, ERp60
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1836658