Learn More
Trajan Scientific Hamilton 100l, X-Type Syringe FN GT
Supplier:  Trajan Scientific 100.SYRXG10022S
Encompass_Preferred
Hamilton 100µl Hamilton X-Type Syringe, Coated Fixed Needle, Gas Tight, PTFE, 22gs, Pt. 3, Hamilton Part Number 204452
Catalog No. 50-635-416