Learn More
Trajan Scientific Hamilton 100l, X-Type Syringe FN GT
Supplier:  Trajan Scientific 100.SYRXG100223
Encompass_Preferred
100µl Hamilton X-Type Syringe, Coated Fixed Needle, Gas Tight, PTFE, 22g, Pt. 3; use Plunger PAL.PLGX100; Hamilton Part Number 204400
Catalog No. 50-635-415