Learn More
Trajan Scientific L-MARK Brand 100l Syringe, PE Plunger Tip, 22g, Gold
Supplier:  Trajan Scientific LMK.2620718C
Encompass_Preferred
L-MARK Brand 100l, Gold Syringe, Coated FN, GT, 22gs, NDL Length 51mm, Pt. Style 3, Plunger Tip PE, Single Made in Germany.
Catalog No. 50-635-328