Learn More
Trajan Scientific L-MARK Brand 100l Syringe, FN, GT, Pt. 3, PE Plunger Tip, 22 Gauge
Supplier:  Trajan Scientific LMK.2620720
Encompass_Preferred
L-MARK Brand 100l, Syringe, FN, GT, 22g, NDL Length 51mm, Pt. Style 3, Plunger Tip PE, Single Made in Germany.
Catalog No. 50-635-333