Learn More
Trajan Scientific L-MARK Brand 100l Syringe, 22s, Pt. 3, PTFE
Catalog No. 50635329
Encompass_Preferred
L-MARK Brand 100l, Syringe, FN, GT, 22s, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single Made in Germany.
Catalog No. 50-635-329 Supplier Trajan Scientific Supplier No. LMK.2620719
L-MARK Brand 100l, Syringe, FN, GT, 22s, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single Made in Germany.