Learn More
Abcam Recombinant Anti-FoxJ1 antibody, Clone EPR21874, 100 μL, Liquid, Rabbit, Targets FoxJ1, Reactivity: Human, Mouse, Rat, Validated for IHC-P, Storage: -20°C
Supplier:  Abcam AB235445100UL
Encompass_Preferred
Recombinant Anti-FoxJ1 antibody, Clone EPR21874, 100 μL, Liquid, Rabbit, Targets FoxJ1, Reactivity: Human, Mouse, Rat, Validated for IHC-P, Storage: -20°C
Catalog No. NC2299924