Learn More
Waters Corp Waters Syringe, 100 μL, High Pressure, Sample
Catalog No. NC9534351 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 100 μL high pressure syringe assembly is a replacement component for ACQUITY Sample Managers. Designed for chromatography systems and high-pressure environments, this precision syringe is essential for accurate sample injection and reliable results.
  • Precision syringe designed for use with chromatography systems.
  • Constructed with durable materials.
  • Ideal for use in high-pressure environments.
  • Essential tool for accurate and precise sample injection.
  • Allows users to achieve reliable and repeatable results.
  • Designed with a highly polished and precisely honed stainless-steel plunger to ensure a smooth, accurate sample delivery.
  • Equipped with a highly sensitive plunger force gauge, which provides users with immediate feedback on the force required for sample injection.
  • Reliable performance and precise accuracy.
Catalog No. NC9534351 Supplier Waters Corp Supplier No. 700002570
This 100 μL high pressure syringe assembly is a replacement component for ACQUITY Sample Managers. Designed for chromatography systems and high-pressure environments, this precision syringe is essential for accurate sample injection and reliable results.
  • Precision syringe designed for use with chromatography systems.
  • Constructed with durable materials.
  • Ideal for use in high-pressure environments.
  • Essential tool for accurate and precise sample injection.
  • Allows users to achieve reliable and repeatable results.
  • Designed with a highly polished and precisely honed stainless-steel plunger to ensure a smooth, accurate sample delivery.
  • Equipped with a highly sensitive plunger force gauge, which provides users with immediate feedback on the force required for sample injection.
  • Reliable performance and precise accuracy.