Learn More
Azzota Corp 10mm Pathlength (0.1Ml) Sub-Micro Cuvette, Pathlength 10mm Inside Width, 2X5mm Volume - 0.1Ml Dimensions - 45X12.5X12.5mm Have Black Walls Z-Height - 15mm, Quartz (190Nm-2500Nm)
Catalog No. 507532877
Encompass_Preferred
10mm Pathlength (0.1ml) Sub-micro Cuvettes Pathlength - 10mm Inside width - 2x5mm Volume - 0.1ml Dimensions - 45x12.5x12.5mm Have black walls Z-Height - 15mm (Option: Z=8.5mm is available) Quartz (190nm-2500nm)
Catalog No. 50-753-2877 Supplier Azzota Corp Supplier No. Q674
10mm Pathlength (0.1ml) Sub-micro Cuvettes Pathlength - 10mm Inside width - 2x5mm Volume - 0.1ml Dimensions - 45x12.5x12.5mm Have black walls Z-Height - 15mm (Option: Z=8.5mm is available) Quartz (190nm-2500nm)