Learn More
Waters Corp Waters Syringe, 100 μL, Rheodyne, 22 Gauge, Square Tip
Catalog No. 50818900 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 100μL Rheodyne Syringe is a low-pressure microsyringe designed for HPLC systems. It features a 2 in. length, 22 Gauge, and a square end (90°) tip style, making it suitable for precise liquid handling.
  • Low-pressure microsyringe
  • Designed for HPLC system type
  • 100 μL volume
  • 2 in. length
  • 22 gauge
  • Square end (90°) tip style
  • Replacement as required
Catalog No. 50-818-900 Supplier Waters Corp Supplier No. WAT033383
The 100μL Rheodyne Syringe is a low-pressure microsyringe designed for HPLC systems. It features a 2 in. length, 22 Gauge, and a square end (90°) tip style, making it suitable for precise liquid handling.
  • Low-pressure microsyringe
  • Designed for HPLC system type
  • 100 μL volume
  • 2 in. length
  • 22 gauge
  • Square end (90°) tip style
  • Replacement as required