Learn More
Genesee Scientific Corporation 100UM NITEX NYLON MESH 40IN W
SDP
Catalog No. NC9357919
Encompass
Ideal For Making Co2 Pads, This Nitex Nylon Mesh Has A 100�m Pore Size And 44% Open Area. Sold By The Contiguous Lineal Foot From A 40In. Roll.
Catalog No. NC9357919 Supplier Genesee Scientific Corporation Supplier No. 57103
Add to Cart
Add to Cart
Ideal For Making Co2 Pads, This Nitex Nylon Mesh Has A 100�m Pore Size And 44% Open Area. Sold By The Contiguous Lineal Foot From A 40In. Roll.