Learn More
Chondrex Inc ChonBlock Detection Antibody Dilution Buffer, 100ml
Catalog No. 501526972
Encompass_Preferred
100 mL of a mammalian protein-based ELISA buffer for dilution of detection/secondary antibody solutions. Please visit www.chondrex.com or email Chondrex, Inc. at support@chondrex.com for more information on how ChonBlock can improve your ELISA results. pH 7.8. Contains 0.4% Tween-20 and 0.04% ProClin 300.
Catalog No. 50-152-6972 Supplier Chondrex Inc Supplier No. 90681
100 mL of a mammalian protein-based ELISA buffer for dilution of detection/secondary antibody solutions. Please visit www.chondrex.com or email Chondrex, Inc. at support@chondrex.com for more information on how ChonBlock can improve your ELISA results. pH 7.8. Contains 0.4% Tween-20 and 0.04% ProClin 300.