Learn More
Chondrex Inc ChonBlock Blocking/Sample Dilution ELISA Buffer 100mL
Catalog No. 501526971
Encompass_Preferred
100 mL of a mammalian protein-based ELISA buffer for ELISA plate blocking or ELISA sample dilution that eliminates background noise reactions caused by non-specific reactions of antibodies in serum/plasma samples. Please visit www.chondrex.com or email Chondrex, Inc. at support@chondrex.com for more information on how ChonBlock can improve your ELISA results. pH 7.8. Contains 0.4% Tween-20 and 0.04% ProClin 300.
Catalog No. 50-152-6971 Supplier Chondrex Inc Supplier No. 9068
100 mL of a mammalian protein-based ELISA buffer for ELISA plate blocking or ELISA sample dilution that eliminates background noise reactions caused by non-specific reactions of antibodies in serum/plasma samples. Please visit www.chondrex.com or email Chondrex, Inc. at support@chondrex.com for more information on how ChonBlock can improve your ELISA results. pH 7.8. Contains 0.4% Tween-20 and 0.04% ProClin 300.