Learn More
Teknova 100X Glucose, Glycerol, IPTG Induction Supplement. 100mL, Sterile. 10 pack.
SDP
Catalog No. 502387501
Encompass
50% Glycerol, 10% Cerelose Anhydrous Dextrose (Glucose),10mM Isopropyl -D-1-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7501 Supplier Teknova Supplier No. G056010PK
50% Glycerol, 10% Cerelose Anhydrous Dextrose (Glucose),10mM Isopropyl -D-1-thiogalactopyranoside(IPTG)