Learn More
ILEX LIFE SCIENCES LLC 100X NEUROPAN-2 10 ML
SDP
Catalog No. 502733711
Encompass_Preferred
PAN-Biotech Neuropan-2 Supplement (100x): Serum-Free Supplement for Cultivation of Neurons (10 ml). DESCRIPTION: Neuropan-2 is a supplement formulated to meet special requirements of neuronal cells. When supplemented with Neuropan Basal Medium, it allows long-term maintenance of normal phenotype and growth of neuronal cells, and maintains pure populations of neuronal cells without need for an astrocyte feeder. Neuropan-2 meets the special cell culture requirements of prenatal and embryonic neuronal cells from hippocampus, cortex, and other regions of the brain. APPLICATION: Neuropan-2 Supplement is a chemically-defined, serum-free supplement for the growth of primary embryonic neurons as well as embryonic neurospheres/CNS progenitor cells when combined with bFGF and EGF in Neuropan Basal Medium. COMPOSTION: Neuropan-2 Supplement contains insulin, transferrin, albumin, and hormones. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3711 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0711010
PAN-Biotech Neuropan-2 Supplement (100x): Serum-Free Supplement for Cultivation of Neurons (10 ml). DESCRIPTION: Neuropan-2 is a supplement formulated to meet special requirements of neuronal cells. When supplemented with Neuropan Basal Medium, it allows long-term maintenance of normal phenotype and growth of neuronal cells, and maintains pure populations of neuronal cells without need for an astrocyte feeder. Neuropan-2 meets the special cell culture requirements of prenatal and embryonic neuronal cells from hippocampus, cortex, and other regions of the brain. APPLICATION: Neuropan-2 Supplement is a chemically-defined, serum-free supplement for the growth of primary embryonic neurons as well as embryonic neurospheres/CNS progenitor cells when combined with bFGF and EGF in Neuropan Basal Medium. COMPOSTION: Neuropan-2 Supplement contains insulin, transferrin, albumin, and hormones. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).