Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free 100X in DMSO) 100x1ml
SDP
Catalog No. 503107270
Encompass_Preferred
Crude cell extracts contain a variety of endogenous enzymes capable of degrading proteins such as phosphatases and proteases The optimal method to enhance the yield of intact proteins is to supplement with inhibitors targeting these known enzymes This Protease Inhibitor Cocktail is used in plant cell and tissue extracts to improve protein stability The cocktail inhibits proteases that degrade both non-phosphorylated and phosphorylated protein substrates The Protease Inhibitor Cocktail contains individual components including AEBSF 1 10-Phenanthroline Bestatin E-64 Leupeptin and Pepstatin A providing broad specificity against cysteine serine aspartic metalloproteases and aminopeptidases This protease inhibitor cocktail has been optimized and tested specifically for plant cell and tissue extracts Provided as a ready-to-use solution this protease inhibitor cocktail is supplied in DMSO as the solvent
Catalog No. 50-310-7270 Supplier Apexbio Technology LLC Supplier No. K1011100
Crude cell extracts contain a variety of endogenous enzymes capable of degrading proteins such as phosphatases and proteases The optimal method to enhance the yield of intact proteins is to supplement with inhibitors targeting these known enzymes This Protease Inhibitor Cocktail is used in plant cell and tissue extracts to improve protein stability The cocktail inhibits proteases that degrade both non-phosphorylated and phosphorylated protein substrates The Protease Inhibitor Cocktail contains individual components including AEBSF 1 10-Phenanthroline Bestatin E-64 Leupeptin and Pepstatin A providing broad specificity against cysteine serine aspartic metalloproteases and aminopeptidases This protease inhibitor cocktail has been optimized and tested specifically for plant cell and tissue extracts Provided as a ready-to-use solution this protease inhibitor cocktail is supplied in DMSO as the solvent