Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free 100X in DMSO) 100x1ml
SDP
Catalog No. 503107437
Encompass_Preferred
Cell crude extracts contain various endogenous enzymes such as phosphatases and proteases capable of degrading proteins present in the extracts The optimal strategy to enhance intact protein yield is adding specific inhibitors targeting these enzymes This Protease Inhibitor Cocktail is used in fungal and yeast extracts to improve protein stability by inhibiting proteases responsible for degrading both phosphorylated and non-phosphorylated protein substrates This Protease Inhibitor Cocktail consists of individual compounds including AEBSF 1 10-Phenanthroline E-64 and Pepstatin A which provide broad-spectrum specific inhibition of cysteine proteases serine proteases aspartic proteases and metalloproteases It is optimized and tested for fungal and yeast extracts This cocktail is available as a ready-to-use solution dissolved in DMSO
Catalog No. 50-310-7437 Supplier Apexbio Technology LLC Supplier No. K1009100
Add to Cart
Add to Cart
Cell crude extracts contain various endogenous enzymes such as phosphatases and proteases capable of degrading proteins present in the extracts The optimal strategy to enhance intact protein yield is adding specific inhibitors targeting these enzymes This Protease Inhibitor Cocktail is used in fungal and yeast extracts to improve protein stability by inhibiting proteases responsible for degrading both phosphorylated and non-phosphorylated protein substrates This Protease Inhibitor Cocktail consists of individual compounds including AEBSF 1 10-Phenanthroline E-64 and Pepstatin A which provide broad-spectrum specific inhibition of cysteine proteases serine proteases aspartic proteases and metalloproteases It is optimized and tested for fungal and yeast extracts This cocktail is available as a ready-to-use solution dissolved in DMSO