Learn More
Ubpbio 100X RELAYTR Europium/Cy5 Ubiquitin Mix, 200 x 20 µl Reactions
Catalog No. NC4036147
Encompass_Preferred
Time resolved (TR)-FRET offers a sensitive, low background, stable and homogenous assay for determination of protein ubiquitination. Europium-ubiquitin and Cy5-ubiquitin are a pair of modified ubiquitin suitable for TR-FRET assays to assess formation of polyubiquitin chains, thereby reporting the activity of E2 and E3 enzymes in the reaction. The concentration and molar ratio of europium-ubiquitin and Cy5-ubiquitin in the RELAYTR Europium/Cy5 Ubiquitin Mix are optimized to achieve excellent signal-to-background ratio in ubiquitination reactions. Optimization of protein concentrations is often necessary to achieve a desirable signal-to-background ratio. Typical protein concentration ranges are: UbE1 (10-30 nM), E2 (25-250 nM) and E3 (10-250 nM).Reaction time is usually 1-3 hours in kinetic or end point assay.
Catalog No. NC4036147 Supplier Ubpbio Supplier No. T3501200
Time resolved (TR)-FRET offers a sensitive, low background, stable and homogenous assay for determination of protein ubiquitination. Europium-ubiquitin and Cy5-ubiquitin are a pair of modified ubiquitin suitable for TR-FRET assays to assess formation of polyubiquitin chains, thereby reporting the activity of E2 and E3 enzymes in the reaction. The concentration and molar ratio of europium-ubiquitin and Cy5-ubiquitin in the RELAYTR Europium/Cy5 Ubiquitin Mix are optimized to achieve excellent signal-to-background ratio in ubiquitination reactions. Optimization of protein concentrations is often necessary to achieve a desirable signal-to-background ratio. Typical protein concentration ranges are: UbE1 (10-30 nM), E2 (25-250 nM) and E3 (10-250 nM).Reaction time is usually 1-3 hours in kinetic or end point assay.