Learn More
CELLTREAT Scientific Products 100mm x 15mm Petri Dish, 3 Compartments, Sterile
SDP
Catalog No. 50828747
Encompass_Preferred
CELLTREAT Scientific Products 3 Compartment Petri Dish. Designed for simultaneous studies of organisms in a common environment. Molded dividers create completely separate compartments while allowing airflow. Ridge on bottom dish allows easy handling and stacking. Lids contain molded spacers to allow gas exchange. Three compartments with working volume of 5mL and growth area of 18.9cm¿. Lid contains stacking rings for increased stability. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.
Catalog No. 50-828-747 Supplier CELLTREAT Scientific Products Supplier No. 229683
CELLTREAT Scientific Products 3 Compartment Petri Dish. Designed for simultaneous studies of organisms in a common environment. Molded dividers create completely separate compartments while allowing airflow. Ridge on bottom dish allows easy handling and stacking. Lids contain molded spacers to allow gas exchange. Three compartments with working volume of 5mL and growth area of 18.9cm¿. Lid contains stacking rings for increased stability. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.