Learn More
CELLTREAT Scientific Products 100mm x 15mm Petri Dish, 4 Compartments, Sterile
SDP
Catalog No. 50828748
Encompass_Preferred
CELLTREAT Scientific Products 4 Compartment Petri Dish. Designed for simultaneous studies of organisms in a common environment. Molded dividers create completely separate compartments while allowing airflow. Ridge on bottom dish allows easy handling and stacking. Lids contain molded spacers to allow gas exchange. Four compartments with working volume of 4mL and growth area of 14.1cm¿. Lid contains stacking rings for increased stability. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.
Catalog No. 50-828-748 Supplier CELLTREAT Scientific Products Supplier No. 229684
CELLTREAT Scientific Products 4 Compartment Petri Dish. Designed for simultaneous studies of organisms in a common environment. Molded dividers create completely separate compartments while allowing airflow. Ridge on bottom dish allows easy handling and stacking. Lids contain molded spacers to allow gas exchange. Four compartments with working volume of 4mL and growth area of 14.1cm¿. Lid contains stacking rings for increased stability. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.