Learn More
CELLTREAT Scientific Products 100mm x 15mm Petri Dish, Slideable, Sterile
SDP
Catalog No. 50969832
Encompass_Preferred
CELLTREAT Scientific Products 100mm x 15mm Slideable Petri Dish. Non-Treated Petri Dish. CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation. Consistent flatness allows for even media distribution. Features partial edge ring on bas wich allows a single dish to be slid from a large stack and enhances stability when stacked. Working volume of 15-16mL and growth area of 55cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.
Catalog No. 50-969-832 Supplier CELLTREAT Scientific Products Supplier No. 229694
CELLTREAT Scientific Products 100mm x 15mm Slideable Petri Dish. Non-Treated Petri Dish. CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation. Consistent flatness allows for even media distribution. Features partial edge ring on bas wich allows a single dish to be slid from a large stack and enhances stability when stacked. Working volume of 15-16mL and growth area of 55cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.