Learn More
CELLTREAT Scientific Products 100mm x 20mm Tissue Culture Treated Dish, Sterile
SDP
Catalog No. 501149056
Encompass_Preferred
CELLTREAT Scientific Products 100mm x 20mm Tissue Culture Treated Dish. Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 15-16mL and growth area of 57cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 300/case.
Catalog No. 50-114-9056 Supplier CELLTREAT Scientific Products Supplier No. 229621
CELLTREAT Scientific Products 100mm x 20mm Tissue Culture Treated Dish. Dishes have thicker and more durable plastic than Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 15-16mL and growth area of 57cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 300/case.