Learn More
Sino Biological Recombinant Human CD84 Protein (His Tag)
Supplier:  Sino Biological 10100H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gly 225) of human CD84 (NP_003865.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3995