Learn More
Sino Biological Recombinant Anti-CD84 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10100R006100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD84 extracellular domain (rh CD84; Catalog#10100-H08H; Met 1-Gly 225; NP_003865.1).
Catalog No. 50-161-2615