Learn More
Sino Biological Anti-NIPP1/Ppp1r8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101001T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human NIPP1/PPP1R8, and purified by antigen affinity chromatography.
Catalog No. 50-161-2587