Learn More
Sino Biological Anti-Syntaxin 6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101005T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Syntaxin 6 / STX6, and purified by antigen affinity chromatography.
Catalog No. 50-161-2593