Learn More
Sino Biological Anti-Syntaxin 8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101006T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human STX8 / Syntaxin 8, and purified by antigen affinity chromatography.
Catalog No. 50-161-2596