Learn More
Sino Biological Anti-TLK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101007T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TLK1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2599