Learn More
Sino Biological Recombinant Human CD55/DAF Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10101H02H100
Encompass_Preferred
A DNA sequence encoding the human CD55 precursor (NP_000565.1) (Met 1-Ser 353) was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-3997