Learn More
Sino Biological Anti-CD55/DAF Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10101MM04A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD55/DAF (rh CD55/DAF; Catalog#10101-H08H; NP_000565.1; Met1-Ser353) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-2667