Learn More
Sino Biological Anti-CD55/DAF Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10101MM04P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD55 / DAF (rh CD55 / DAF; Catalog#10101-H08H; NP_000565.1; Met1-Ser353) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-2672