Learn More
Sino Biological Recombinant Anti-CD55/DAF Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 10101R028C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD55 / DAF (rh CD55 / DAF; Catalog#10101-H08H; NP_000565.1; Met 1-Ser 353) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-161-2680