Learn More
Sino Biological Anti-CD55/DAF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10101RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD55 / DAF (rh CD55 / DAF; Catalog#10101-H08H; NP_000565.1; Met1-Ser353). CD55 / DAF specific IgG was purified by Human CD55 / DAF affinity chromatography.
Catalog No. 50-161-2688