Learn More
Sino Biological Anti-TRIM37 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101014T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TRIM37, and purified by antigen affinity chromatography.
Catalog No. 50-161-2638