Learn More
Sino Biological Anti-UBE4A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101019T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human UBE4A, and purified by antigen affinity chromatography.
Catalog No. 50-161-2656